2-Bromoanisole 97%

Supplier: Thermo Fisher Scientific
Warning

170260250 170261000
ACRO170260250EA 72.9 EUR
ACRO170260250 ACRO170261000
2-Bromoanisole 97%
2-Bromoanisole
Formula: C₇H₇BrO
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1,502 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00000064
CAS Number: 578-57-4
EINECS: 209-425-8
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5730 to 1.5750 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR