3-Bromoanisole 99+%
Supplier: Thermo Fisher Scientific
|
|
Synonyms:
1-Bromo-3-methoxybenzene
106620250
106621000
ACRO106620250EA
59
EUR
ACRO106620250
ACRO106621000
3-Bromoanisole 99+%
3-Bromoanisole
Formula:
C₇H₇BrO Boiling Pt: 211 °C (1013 hPa) Melting Pt: 2 °C Density: 1,47 g/cm³ Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000081 CAS Number: 2398-37-0 EINECS: 219-264-5 |
Specification Test Results
Appearance | Clear colorless to yellow-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5630 to 1.5650 (20°C, 589 nm) |
Water | ≤0.1 % |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...