4-Bromoanisole ≥99%

Supplier: Thermo Fisher Scientific
Warning

A11824.0E A11824.22 A11824.36
A11824.0EEA 371 EUR
A11824.0E A11824.22 A11824.36
4-Bromoanisole ≥99%
4-Bromoanisole

Purity: 99% CAS No.: 104-92-7 Molecular Formula: C7H7BrO Formula Weight: 187.04 Flash Point: 94°(201°F) Physical Form: Liquid Density: 1.494 Refractive Index: 1.5640 Melting Point: 10-13° Boiling Point: 98-99°/10mm Beilstein Reference: 1237590 Merck Reference: 14,1427 MDL No.: MFCD00000097

Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 98…99 °C (10 mmHg)
Melting Pt: 10…13 °C
Density: 1.494
Flash Pt: 94 °C (201 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000097
CAS Number: 104-92-7
EINECS: 203-252-1
Merck Index: 13,01412

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR