4-Bromoveratrole 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-Bromo-3,4-dimethoxybenzene, 4-Bromo-1,2-dimethoxybenzene
107540250
107541000
107540050
ACRO107540250EA
75.2
EUR
ACRO107540250
ACRO107541000
ACRO107540050
4-Bromoveratrole 97%
4-Bromoveratrole
Formula:
C₈H₉BrO₂ MW: 217,06 g/mol Boiling Pt: 255 °C (1013 hPa) Density: 1,5 g/cm³ Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008381 CAS Number: 2859-78-1 EINECS: 220-677-8 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5705 to 1.5755 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...