2-Bromophenol 98%

Supplier: Thermo Fisher Scientific
Warning

157360100 157360500
ACRO157360100EA 73.7 EUR
ACRO157360100 ACRO157360500
2-Bromophenol 98%
2-Bromophenol
Formula: C₆H₅BrO
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: 5 °C
Density: 1,49 g/cm³
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00002146
CAS Number: 95-56-7
EINECS: 202-432-7
UN: 1993
ADR: 3,III
Merck Index: 13,01413

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5880 to 1.5900 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR