2-Bromophenol ≥98%

Supplier: Thermo Fisher Scientific
Warning

A14112.14 A14112.22 A14112.36
A14112.14EA 126 EUR
A14112.14 A14112.22 A14112.36
2-Bromophenol ≥98%
2-Bromophenol

Purity: 98% CAS No.: 95-56-7 Molecular Formula: C6H5BrO Formula Weight: 173.01 Flash Point: 89°(192°F) Refractive Index: 1.5890 Odour: Phenol-like Melting Point: 5° Boiling Point: 194-195° Beilstein Reference: 1905115 Merck Reference: 14,1428 UN No.: 3082 MDL No.: MFCD00002146

Formula: C₆H₅BrO
MW: 173.01 g/mol
Boiling Pt: 194…195 °C
Melting Pt: 5 °C
Density: 1.636
Flash Pt: 92 °C (192 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002146
CAS Number: 95-56-7
EINECS: 202-432-7
UN: 3082
ADR: 9 III
Merck Index: 13,01413

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR