3-Bromophenol ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 98%
CAS No.: 591-20-8
Molecular Formula: C6H5BrO
Formula Weight: 173.01
Flash Point:
Formula:
C₆H₅BrO Boiling Pt: 235…236 °C Melting Pt: 28…32 °C Density: 1,63 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002253 CAS Number: 591-20-8 EINECS: 209-706-5 UN: 2811 ADR: 6.1,III Merck Index: 14,1428 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...