3-Nitrobenzaldehyde 99%
Supplier: Thermo Scientific
|
Warning
|
128411000
128415000
ACRO128411000EA
34.8
EUR
ACRO128411000
ACRO128415000
3-Nitrobenzaldehyde 99%
3-Nitrobenzaldehyde
Formula:
C₇H₅NO₃ Boiling Pt: 232 °C (1013 hPa) Melting Pt: 56…59 °C Density: 1,481 g/cm³ (0 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007249 CAS Number: 99-61-6 EINECS: 202-772-6 UN: 3077 Merck Index: 12,06684 |
Specification Test Results
Appearance | Yellow to yellow-brown Granular powder |
Infrared spectrum | Conforms |
Melting point | 56 to 59 °C |
Water | ≤0.5 % |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...