4-Nitrobenzaldehyde 99%

Supplier: Thermo Fisher Scientific
Warning

156990250 156991000 156995000
ACRO156990250EA 49 EUR
ACRO156990250 ACRO156991000 ACRO156995000
4-Nitrobenzaldehyde 99%
4-Nitrobenzaldehyde
Formula: C₇H₅NO₃
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 105…106 °C
Density: 1,546 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007346
CAS Number: 555-16-8
EINECS: 209-084-5
Merck Index: 12,06684

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR