5-Hydroxy-2-nitrobenzaldehyde ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
L12051.03
L12051.06
L12051.03EA
0
EUR
L12051.03
L12051.06
5-Hydroxy-2-nitrobenzaldehyde ≥97%
5-Hydroxy-2-nitrobenzaldehyde
Purity: 97%
CAS No.: 42454-06-8
Molecular Formula: C7H5NO4
Formula Weight: 167.12
Melting Point: 166-168°
Beilstein Reference: 1910196
Formula:
C₇H₅NO₄ Melting Pt: 166…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007332 CAS Number: 42454-06-8 EINECS: 255-832-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...