Salicylaldehyde ethyl ether ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Ethoxybenzaldehyde

A15035.18 A15035.22
A15035.18EA 47 EUR
A15035.18 A15035.22
Salicylaldehyde ethyl ether ≥97%
Salicylaldehyde ethyl ether

Purity: 97+% CAS No.: 613-69-4 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 91°(195°F) Refractive Index: 1.5430 Boiling Point: 249° Beilstein Reference: 1937270 MDL No.: MFCD00003316

Formula: C₉H₁₀O₂
MW: 150,18 g/mol
Boiling Pt: 249 °C
Melting Pt: 20…22 °C
Density: 1,077
Flash Pt: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003316
CAS Number: 613-69-4
EINECS: 210-349-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR