3,5-Dimethoxybenzaldehyde 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
155220100
155220500
ACRO155220100EA
113
EUR
ACRO155220100
ACRO155220500
3,5-Dimethoxybenzaldehyde 98%
3,5-Dimethoxybenzaldehyde
Formula:
C₉H₁₀O₃ Boiling Pt: 151 °C (16 mmHg) Melting Pt: 44…48 °C Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003366 CAS Number: 7311-34-4 EINECS: 230-772-6 |
Specification Test Results
Appearance | White to beige Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 44 to 48 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...