3,4-Dihydroxybenzaldehyde for synthesis, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

8.20475.0025 8.20475.0100
8.20475.0025EA 121 EUR
8.20475.0025 8.20475.0100
3,4-Dihydroxybenzaldehyde for synthesis, Sigma-Aldrich®
3,4-Dihydroxybenzaldehyde
Formula: C₇H₆O₃
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 150…155 °C
Density: 1,505 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003370
CAS Number: 139-85-5
EINECS: 205-377-7
Merck Index: 12,08079

Order Now

Specification Test Results

Solubility 6.3 g/l (25 °C)
Melting Point 152 - 156 °C
Molar Mass 138.12 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR