Anthranilonitrile 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A12286.18
A12286.30
A12286.18EA
50.2
EUR
A12286.18
A12286.30
Anthranilonitrile 98%
Anthranilonitrile
Purity: 98%
CAS No.: 1885-29-6
Molecular Formula: C7H6N2
Formula Weight: 118.14
Flash Point: 145°(293°F)
Odour: Almond-like
Boiling Point: 286°
Beilstein Reference: 907187
UN No.: 3439
MDL No.: MFCD00007631
Formula:
C₇H₆N₂ Boiling Pt: 286 °C Melting Pt: 45…49 °C Density: 1,26 g/cm³ (20 °C) Flash Pt: 145 °C (293 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007631 CAS Number: 1885-29-6 EINECS: 217-549-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...