Silver benzoate, hydrate

Supplier: Thermo Scientific

Synonyms: Benzoic acid silver (I) salt

11896.06 11896.14 11896.22
11896.06EA 38 EUR
11896.06 11896.14 11896.22
Silver benzoate, hydrate
Silver benzoate

CAS No.: 532-31-0 Molecular Formula: C7H5AgO2g.xH2O Formula Weight: 228.89(anhy) Physical Form: Powder Sensitivity: Light Sensitive Appearance: White to cream or grey MDL No.: MFCD00013030

Formula: C₇H₅AgO₂
MW: 228,98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013030
CAS Number: 532-31-0
EINECS: 208-533-2
Merck Index: 13,01092

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR