5-Nitroanthranilic acid 98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Amino-5-nitrobenzoic acid

A10426.09 A10426.18
A10426.09EA 71.4 EUR
A10426.09 A10426.18
5-Nitroanthranilic acid 98%
5-Nitroanthranilic acid

Purity: 98% CAS No.: 616-79-5 Molecular Formula: C7H6N2O4 Formula Weight: 182.14 Melting Point: ca 270° dec. Beilstein Reference: 646219

Formula: C₇H₆N₂O₄
MW: 182,14 g/mol
Melting Pt: ∼270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00017039
CAS Number: 616-79-5
EINECS: 210-493-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR