4,5-Dimethoxyanthranilic acid 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-Amino-4,5-dimethoxybenzoic acid, 6-Aminoveratric acid
L06205.14
L06205.06
L06205.14EA
82.5
EUR
L06205.14
L06205.06
4,5-Dimethoxyanthranilic acid 98%
4,5-Dimethoxyanthranilic acid
Purity: 98%
CAS No.: 5653-40-7
Molecular Formula: C9H11NO4
Formula Weight: 197.19
Melting Point: ca 155-158° dec.
Beilstein Reference: 782814
Formula:
C₉H₁₁NO₄ MW: 197.19 g/mol Boiling Pt: 359 Melting Pt: ∼155…158 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011671 CAS Number: 5653-40-7 EINECS: 227-095-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...