2-Nitrobenzoic acid 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
278320050
278322500
278320010
278320250
ACRO278320050EA
31
EUR
ACRO278320050
ACRO278322500
ACRO278320010
ACRO278320250
2-Nitrobenzoic acid 95%
2-Nitrobenzoic acid
Formula:
C₇H₅NO₄ Melting Pt: 141…148 °C Density: 1,575 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007137 CAS Number: 552-16-9 EINECS: 209-004-9 Merck Index: 12,06686 |
Specification Test Results
Appearance | White to cream to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 143 to 149 °C |
HPLC | ≥94 % |
Titration with NaOH | 94 to 106 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...