5-Hydroxy-2-nitrobenzoic acid ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
H29194.03
H29194.09
H29194.03EA
44.6
EUR
H29194.03
H29194.09
5-Hydroxy-2-nitrobenzoic acid ≥99%
5-Hydroxy-2-nitrobenzoic acid
Purity: 99%
CAS No.: 610-37-7
Molecular Formula: C7H5NO5
Formula Weight: 183.12
Melting Point: 168-172°
MDL No.: MFCD00017566
Formula:
C₇H₅NO₅ Melting Pt: 168…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017566 CAS Number: 610-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...