o-Anisic acid 99%

Supplier: Thermo Fisher Scientific
Warning

173751000 173755000 173750025
ACRO173751000EA 53.3 EUR
ACRO173751000 ACRO173755000 ACRO173750025
o-Anisic acid 99%
o-Anisic acid
Formula: C₈H₈O₃
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 98…102 °C
Density: 1,38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002431
CAS Number: 579-75-9
EINECS: 209-447-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 102 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR