4-Propoxybenzoic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

B24958.06 B24958.14
B24958.06EA 33 EUR
B24958.06 B24958.14
4-Propoxybenzoic acid ≥98%
4-Propoxybenzoic acid

Purity: 98% CAS No.: 5438-19-7 Molecular Formula: C10H12O3 Formula Weight: 180.20 Melting Point: 145-147° Beilstein Reference: 2208387

Formula: C₁₀H₁₂O₃
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 145…147 °C
Density: 1,278 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013989
CAS Number: 5438-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR