3-Methoxysalicylic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Hydroxy-3-methoxybenzoic acid
L01658.06
L01658.14
L01658.06EA
25.6
EUR
L01658.06
L01658.14
3-Methoxysalicylic acid ≥98%
3-Methoxysalicylic acid
Purity: 98+%
CAS No.: 877-22-5
Molecular Formula: C8H8O4
Formula Weight: 168.15
Melting Point: 151-152°
Beilstein Reference: 2209642
Formula:
C₈H₈O₄ MW: 168,15 g/mol Melting Pt: 151…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002445 CAS Number: 877-22-5 EINECS: 212-888-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...