4-Methoxysalicylic acid 99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Hydroxy-p-anisic acid, 2-Hydroxy-4-methoxybenzoic acid
174240100
174240500
ACRO174240100EA
78.7
EUR
ACRO174240100
ACRO174240500
4-Methoxysalicylic acid 99%
4-Methoxysalicylic acid
Formula:
C₈H₈O₄ MW: 168,15 g/mol Boiling Pt: 327 °C (1013 hPa) Melting Pt: 157…159 °C |
MDL Number:
MFCD00002450 CAS Number: 2237-36-7 EINECS: 218-801-0 |
Specification Test Results
Appearance | Off-white to beige Powder |
Infrared spectrum | Conforms |
Melting point | 157 to 161 °C |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...