2,3,4-Trimethoxybenzoic acid 98+%

Supplier: Thermo Fisher Scientific
Warning

183470250 183471000
ACRO183470250EA 39.3 EUR
ACRO183470250 ACRO183471000
2,3,4-Trimethoxybenzoic acid 98+%
2,3,4-Trimethoxybenzoic acid
Formula: C₁₀H₁₂O₅
Boiling Pt: 110 - 115 °C (0,04 -rr)
Melting Pt: 99…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00002433
CAS Number: 573-11-5
EINECS: 209-350-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 99 to 102 °C
Titration with NaOH ≥98.0 %
Residue after ignition ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR