Chlorobenzene ≥99.9% for spectrophotometry

Supplier: Thermo Scientific
Warning

Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene

22921.AP 22921.K2 22921.M1
22921.APEA 93.3 EUR
22921.AP 22921.K2 22921.M1
Chlorobenzene ≥99.9% for spectrophotometry
Chlorobenzene

Purity: 99.9% CAS No.: 108-90-7 Molecular Formula: C6H5Cl Formula Weight: 112.56 Flash Point: 28°(82°F) Physical Form: Liquid Density: 1.106 Refractive Index: 1.5240 Appearance: Colourless Odour: Aromatic Melting Point: -45° Boiling Point: 132° Beilstein Reference: 605632 Merck Reference: 14,2121 UN No.: 1134 MDL No.: MFCD00000530

Formula: C₆H₅Cl
MW: 112,56 g/mol
Boiling Pt: 132 °C
Melting Pt: –45 °C
Density: 1,106
Flash Pt: 28 °C (82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45
Merck Index: 14,2121
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR