2-Chloro-5-nitrobenzaldehyde ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
A14824.09
A14824.18
A14824.09EA
40.5
EUR
A14824.09
A14824.18
2-Chloro-5-nitrobenzaldehyde ≥97%
2-Chloro-5-nitrobenzaldehyde
Purity: 97%
CAS No.: 6361-21-3
Molecular Formula: C7H4ClNO3
Formula Weight: 185.57
Melting Point: 77-81°
Beilstein Reference: 743764
Formula:
C₇H₄ClNO₃ Melting Pt: 77…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007293 CAS Number: 6361-21-3 EINECS: 228-830-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...