3-Chlorobenzonitrile ≥99%

Supplier: Thermo Scientific
Danger

Synonyms: m-Chlorobenzonitrile

A13237.09 A13237.18 A13237.30
A13237.09EA 38.2 EUR
A13237.09 A13237.18 A13237.30
3-Chlorobenzonitrile ≥99%
3-Chlorobenzonitrile

Purity: 99% CAS No.: 766-84-7 Molecular Formula: C7H4ClN Formula Weight: 137.57 Flash Point: 97°(206°F) Odour: Almond-like Boiling Point: 94°/10mm Beilstein Reference: 1906694 UN No.: 3439 MDL No.: MFCD00001798

Formula: C₇H₄ClN
MW: 137,57 g/mol
Boiling Pt: 94 °C (10 mmHg)
Melting Pt: 39…42 °C
Density: 1,14
Flash Pt: 97 °C (206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001798
CAS Number: 766-84-7
EINECS: 212-172-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR