Dichlobenil ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 2,6-Dichlorobenzonitrile

A14986.09 A14986.14 A14986.22
A14986.09EA 23.7 EUR
A14986.09 A14986.14 A14986.22
Dichlobenil ≥98%
Dichlobenil

Purity: 98% CAS No.: 1194-65-6 Molecular Formula: C7H3Cl2N Formula Weight: 172.01 Odour: Aromatic Melting Point: 143-147° Boiling Point: 270° Beilstein Reference: 1909167 Merck Reference: 14,3042 UN No.: 3077 MDL No.: MFCD00001781

Formula: C₇H₃Cl₂N
MW: 172,01 g/mol
Boiling Pt: 270 °C
Melting Pt: 143…147 °C
Density: 1,623 g/cm³ (–101 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001781
CAS Number: 1194-65-6
EINECS: 214-787-5
UN: 3077
ADR: 9,III
Merck Index: 14,3042

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR