2-Amino-5-chlorobenzonitrile 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
5-Chloroanthranilonitrile
B22690.03
B22690.06
B22690.03EA
30.1
EUR
B22690.03
B22690.06
2-Amino-5-chlorobenzonitrile 97%
2-Amino-5-chlorobenzonitrile
Purity: 97%
CAS No.: 5922-60-1
Molecular Formula: C7H5ClN2
Formula Weight: 152.58
Melting Point: 94-97°
Boiling Point: 132-135°/0.5mm
UN No.: 3439
MDL No.: MFCD00017106
Formula:
C₇H₅ClN₂ MW: 152,58 g/mol Boiling Pt: 132…135 °C (0,5 mmHg) Melting Pt: 94…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017106 CAS Number: 5922-60-1 EINECS: 227-651-5 UN: 3439 ADR: 6.1,III |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...