4-Chloroanthranilic acid 98%

Supplier: Thermo Fisher Scientific
Warning

103320250 103321000
ACRO103320250EA 57.5 EUR
ACRO103320250 ACRO103321000
4-Chloroanthranilic acid 98%
4-Chloroanthranilic acid
Formula: C₇H₆ClNO₂
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 231…235 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007778
CAS Number: 89-77-0
EINECS: 201-938-5

Order Now

Specification Test Results

Appearance Beige to light brown Powder
Infrared spectrum Conforms
Melting point 231 to 235 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR