2-Chloro-5-nitrobenzoic acid 99+%
Supplier: Thermo Scientific
|
Warning
|
154691000
154695000
ACRO154691000EA
42.9
EUR
ACRO154691000
ACRO154695000
2-Chloro-5-nitrobenzoic acid 99+%
2-Chloro-5-nitrobenzoic acid
Formula:
C₇H₄ClNO₄ MW: 201,57 g/mol Melting Pt: 164…168 °C Density: 0,4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007294 CAS Number: 2516-96-3 EINECS: 219-739-7 |
Specification Test Results
Appearance | Light yellow to beige to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 164 to 168 °C |
Loss on drying | ≤0.5 % |
HPLC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...