5-Chloro-2-fluoroaniline ≥97%

Supplier: Thermo Fisher Scientific
Warning

B20951.09 B20951.18
B20951.09EA 55.8 EUR
B20951.09 B20951.18
5-Chloro-2-fluoroaniline ≥97%
5-Chloro-2-fluoroaniline

Purity: 97% CAS No.: 2106-05-0 Molecular Formula: C6H5ClFN Formula Weight: 145.56 Flash Point: 103°(217°F) Refractive Index: 1.5600 Odour: Characteristic Boiling Point: 102-105°/20mm UN No.: 2810 MDL No.: MFCD00069416

Formula: C₆H₅ClFN
Boiling Pt: 102…105 °C (20 mmHg)
Density: 1,30
Flash Pt: 103 °C (217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00069416
CAS Number: 2106-05-0
EINECS: 218-284-1
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR