3-Chloro-4-fluorophenol ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
A14382.06
A14382.14
A14382.06EA
31.1
EUR
A14382.06
A14382.14
3-Chloro-4-fluorophenol ≥97%
3-Chloro-4-fluorophenol
Purity: 97%
CAS No.: 2613-23-2
Molecular Formula: C6H4ClFO
Formula Weight: 146.55
Flash Point: 108°(226°F)
Odour: Phenol-like
Boiling Point: 103-104°/11mm
Beilstein Reference: 3236443
UN No.: 3261
MDL No.: MFCD00002257
Formula:
C₆H₄ClFO Boiling Pt: 103…104 °C (11 mmHg) Melting Pt: 38…43 °C Flash Pt: 108 °C (226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002257 CAS Number: 2613-23-2 EINECS: 220-042-5 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...