1-Chloro-2-iodobenzene 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
249460050
249460500
ACRO249460050EA
32.3
EUR
ACRO249460050
ACRO249460500
1-Chloro-2-iodobenzene 99%
1-Chloro-2-iodobenzene
Formula:
C₆H₄ClI Boiling Pt: 234 °C (1013 hPa) Melting Pt: 1 °C Density: 1,95 g/cm³ Flash Pt: 111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001033 CAS Number: 615-41-8 EINECS: 210-423-4 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6330 to 1.6350 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...