3-Chloroaniline 99%
Supplier: Thermo Fisher Scientific
|
Danger
|
108580050
108581000
108585000
ACRO108580050EA
28.5
EUR
ACRO108580050
ACRO108581000
ACRO108585000
3-Chloroaniline 99%
3-Chloroaniline
Formula:
C₆H₆ClN MW: 127.57 g/mol Boiling Pt: 230.5 Melting Pt: –11 °C Density: 1.2 g/cm³ Flash Pt: 118 Storage Temperature: Ambient |
MDL Number:
MFCD00007765 CAS Number: 108-42-9 EINECS: 203-581-0 UN: 2019 ADR: 6.1 II Merck Index: 13,02136 |
Specification Test Results
Appearance | Clear light yellow to brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5927 to 1.5947 (20°C, 589 nm) |
GC | ≥98.5 % |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...