2-Chloro-4,6-dinitroaniline ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: (2-Chloro-4,6-dinitrophenyl)amine, 6-Chloro-2,4-dinitroaniline

B24379.22 B24379.36
B24379.22EA 33.5 EUR
B24379.22 B24379.36
2-Chloro-4,6-dinitroaniline ≥97%
2-Chloro-4,6-dinitroaniline

Purity: 97% CAS No.: 3531-19-9 Molecular Formula: C6H4ClN3O4 Formula Weight: 217.57 Melting Point: 153-155° UN No.: 2811 MDL No.: MFCD00007147

Formula: C₆H₄ClN₃O₄
MW: 217,57 g/mol
Melting Pt: 153…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00007147
CAS Number: 3531-19-9
EINECS: 222-564-9
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR