2,6-Dichlorothiophenol ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2,6-Dichlorobenzenethiol, 2,6-Dichlorobenzene-1-thiol, 1,3-Dichloro-2-mercaptobenzene

A12854.06 A12854.14
A12854.06EA 34.7 EUR
A12854.06 A12854.14
2,6-Dichlorothiophenol ≥97%
2,6-Dichlorothiophenol
Formula: C₆H₄Cl₂S
MW: 179.07 g/mol
Boiling Pt: 101…103 °C (14 mmHg)
Melting Pt: 46…50 °C
Flash Pt: 112 °C (233 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004833
CAS Number: 24966-39-0
EINECS: 416-720-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR