3-Fluorobenzaldehyde ≥97%
Supplier: Thermo Scientific
|
Danger
|
A11396.09
A11396.18
A11396.30
A11396.09EA
34.8
EUR
A11396.09
A11396.18
A11396.30
3-Fluorobenzaldehyde ≥97%
3-Fluorobenzaldehyde
Purity: 97%
CAS No.: 456-48-4
Molecular Formula: C7H5FO
Formula Weight: 124.11
Flash Point: 56°(132°F)
Refractive Index: 1.5190
Boiling Point: 173°
Beilstein Reference: 970178
UN No.: 1989
MDL No.: MFCD00003348
Formula:
C₇H₅FO MW: 124,11 g/mol Boiling Pt: 173 °C Density: 1,174 Flash Pt: 56 °C (132 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003348 CAS Number: 456-48-4 EINECS: 207-266-9 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...