3,5-Difluoroanisole ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1,3-Difluoro-5-methoxybenzene
A15497.14
A15497.14EA
186
EUR
A15497.14
3,5-Difluoroanisole ≥97%
3,5-Difluoroanisole
Purity: 97%
CAS No.: 93343-10-3
Molecular Formula: C7H6F2O
Formula Weight: 144.12
Flash Point: 43°(109°F)
Refractive Index: 1.4660
Boiling Point: 145-148°
Beilstein Reference: 4966844
UN No.: 1993
MDL No.: MFCD00042560
Formula:
C₇H₆F₂O MW: 144,12 g/mol Boiling Pt: 145…148 °C Density: 1,234 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042560 CAS Number: 93343-10-3 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...