4-Fluoroanisole ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-Fluoro-4-methoxybenzene
B23048.14
B23048.22
B23048.14EA
31
EUR
B23048.14
B23048.22
4-Fluoroanisole ≥99%
4-Fluoroanisole
Purity: 99%
CAS No.: 459-60-9
Molecular Formula: C7H7FO
Formula Weight: 126.13
Flash Point: 43°(109°F)
Refractive Index: 1.4880
Melting Point: -45°
Boiling Point: 156-158°
Beilstein Reference: 1281425
UN No.: 1993
MDL No.: MFCD00000348
Formula:
C₇H₇FO MW: 126,13 g/mol Boiling Pt: 156…158 °C Melting Pt: –45 °C Density: 1,116 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000348 CAS Number: 459-60-9 EINECS: 207-295-7 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...