3-Fluorophenol 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
119550100
119550500
119552000
ACRO119550100EA
47.9
EUR
ACRO119550100
ACRO119550500
ACRO119552000
3-Fluorophenol 98%
3-Fluorophenol
Formula:
C₆H₅FO MW: 112.1 g/mol Boiling Pt: 178 °C (760 mmHg) Melting Pt: 10 °C Density: 1.24 g/cm³ Flash Pt: 71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002254 CAS Number: 372-20-3 EINECS: 206-748-6 UN: 2810 ADR: 6.1 III |
Specification Test Results
Appearance | Clear colorless to yellow-brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5125 to 1.5155 (20°C, 589 nm) |
GC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...