Pentafluorophenol ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2,3,4,5,6-Pentafluorophenol

A15574.06 A15574.14 A15574.22
A15574.06EA 43.3 EUR
A15574.06 A15574.14 A15574.22
Pentafluorophenol ≥99%
Pentafluorophenol

Purity: 99% CAS No.: 771-61-9 Molecular Formula: C6HF5O Formula Weight: 184.06 Flash Point: 72°(161°F) Refractive Index: 1.4270 Melting Point: 32-36° Boiling Point: 143° Beilstein Reference: 1912584 MDL No.: MFCD00002156

Formula: C₆HF₅O
MW: 184.07 g/mol
Boiling Pt: 143 °C
Melting Pt: 32…36 °C
Density: 1.757
Flash Pt: 72 °C (161 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002156
CAS Number: 771-61-9
EINECS: 212-235-8
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR