3-Iodophenol 99%
Supplier: Thermo Scientific
|
Warning
|
249700250
249700050
ACRO249700250EA
160
EUR
ACRO249700250
ACRO249700050
3-Iodophenol 99%
3-Iodophenol
Formula:
C₆H₅IO MW: 220,01 g/mol Melting Pt: 38…43 °C Flash Pt: 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002261 CAS Number: 626-02-8 EINECS: 210-923-2 ADR: 9, |
Specification Test Results
Appearance | White to yellow to brown or grey or purple or fused solid |
Infrared spectrum | Conforms |
Melting point | 37 to 43 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...