4-Iodophenol 99%
Supplier: Thermo Scientific
|
Danger
|
122390100
122390250
122391000
ACRO122390100EA
66.5
EUR
ACRO122390100
ACRO122390250
ACRO122391000
4-Iodophenol 99%
4-Iodophenol
Formula:
C₆H₅IO MW: 220,01 g/mol Boiling Pt: 287 °C (760 mmHg) Melting Pt: 89…94 °C Density: 2,13 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002327 CAS Number: 540-38-5 EINECS: 208-745-5 UN: 3261 ADR: 8,III Merck Index: 13,05058 |
Specification Test Results
Appearance | Beige to pink to brown Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 89 to 94 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...