4-Iodophenol ≥98%

Supplier: Thermo Scientific
Danger

A13455.14 A13455.06 A13455.22
A13455.14EA 106 EUR
A13455.14 A13455.06 A13455.22
4-Iodophenol ≥98%
4-Iodophenol

Purity: 98+% CAS No.: 540-38-5 Molecular Formula: C6H5IO Formula Weight: 220.01 Melting Point: 91-95° Boiling Point: 138°/5mm Merck Reference: 14,5036 MDL No.: MFCD00002327

Formula: C₆H₅IO
MW: 220,01 g/mol
Boiling Pt: 138 °C (5 mmHg)
Melting Pt: 91…95 °C
Density: 2,13 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002327
CAS Number: 540-38-5
EINECS: 208-745-5
UN: 3261
ADR: 8,III
Merck Index: 14,5036

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR