3-Nitrophenol 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
m-Nitrophenol
172300100
172300500
172302500
ACRO172300100EA
52.4
EUR
ACRO172300100
ACRO172300500
ACRO172302500
3-Nitrophenol 99%
3-Nitrophenol
Formula:
C₆H₅NO₃ MW: 139,11 g/mol Boiling Pt: 194 °C (70 mmHg) Melting Pt: 94…98 °C Density: 1,49 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00007240 CAS Number: 554-84-7 EINECS: 209-079-8 UN: 1663 ADR: 6.1,III Merck Index: 12,06682 |
Specification Test Results
Appearance | Yellow to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 94 to 98 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...