2-Nitrophenol for synthesis, Sigma-Aldrich®

Supplier: Merck

Synonyms: o-Nitrophenol

M-Clarity™ quality level = MQ200

8.06790.0100 8.06790.0500
8.06790.0100EA 36.1 EUR
8.06790.0100 8.06790.0500
2-Nitrophenol for synthesis, Sigma-Aldrich®
2-Nitrophenol
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 214.6
Melting Pt: 43…45 °C
Density: 1.599 g/cm³ (20 °C)
Flash Pt: 108
Storage Temperature: Ambient
MDL Number: MFCD00011688
CAS Number: 88-75-5
EINECS: 201-857-5
UN: 1663
ADR: 6.1 III
Merck Index: 12,06717

Order Now

Specification Test Results

Solubility 2.1 g/l (20 °C)
Melting Point 43 - 45 °C
Molar Mass 139.11 g/mol
Boiling Point 215 - 216 °C (1013 hPa)
Vapor Pressure 0.15 hPa (25 °C)
Flash Point 108 °C
Ignition Point 550 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR