2-Ethoxyphenol for synthesis, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1-Hydroxy-2-ethoxybenzene, Pyrocatechol monoethyl ether, o-Ethoxyphenol

M-Clarity™ quality level = MQ200

8.01917.0250 8.01917.9050
8.01917.0250EA 49.3 EUR
8.01917.0250 8.01917.9050
2-Ethoxyphenol for synthesis, Sigma-Aldrich®
2-Ethoxyphenol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 214…216 °C (1013 hPa)
Melting Pt: 28…29 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 86
Storage Temperature: Ambient
MDL Number: MFCD00002187
CAS Number: 94-71-3
EINECS: 202-358-5

Order Now

Specification Test Results

Solubility 9 g/l (30 °C)
Melting Point 24 - 28 °C
Molar Mass 138.16 g/mol
Boiling Point 214 - 216 °C (1013 hPa)
Vapor Pressure 1 hPa (45 °C)
Flash Point 86 °C
Refractive Index 1.5291 (25 °C, 589 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR