2-Chloro-3-fluorobenzoic acid 97%

Supplier: Thermo Scientific
Warning

H31761.03 H31761.06
H31761.03EA 0 EUR
H31761.03 H31761.06
2-Chloro-3-fluorobenzoic acid 97%
2-Chloro-3-fluorobenzoic acid

Purity: 97% CAS No.: 102940-86-3 Molecular Formula: C7H4ClFO2 Formula Weight: 174.56 Melting Point: 170-172° MDL No.: MFCD00973903

Formula: C₇H₄ClFO₂
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00973903
CAS Number: 102940-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR