2-Bromo-1-methylimidazole 95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Bromo-1-methyl-1H-imidazole
H52407.03
H52407.06
H52407.03EA
133
EUR
H52407.03
H52407.06
2-Bromo-1-methylimidazole 95%
2-Bromo-1-methylimidazole
Purity: 95%
CAS No.: 16681-59-7
Molecular Formula: C4H5BrN2
Formula Weight: 161.00
Flash Point:
Formula:
C₄H₅BrN₂ MW: 161 g/mol Boiling Pt: 172 °C Density: 1,649 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02179525 CAS Number: 16681-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...